Learn More
Fisherbrand™ Colored Screw Caps
For use with nonsterile skirted- and conical-bottom tubes
Brand:  Fisherbrand™
UNSPSC : 41120000
Code : A0
Additional Details : Weight : 0.14000kg
 View more versions of this product
Product Code. 11920089
  • £34.85 / Pack of 500
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications Description & Specifications
Specifications
Yes
Yellow
Non-sterile skirted and conical bottom tubes
Polypropylene
Screw Cap with O-ring
O-ring