Learn More
Fisherbrand™ Colored Screw Caps
For use with nonsterile skirted- and conical-bottom tubes
Brand:  Fisherbrand™
Code : A0
Additional Details : Weight : 0.15000kg
 View more versions of this product
Product Code. 11900089
  • £36.57 / Pack of 500
Estimated Shipment
11-06-2018
Add to basket
Description
Description
  • Feature a frosted writing surface
  • Can be pierced for sample removal
Specifications
Specifications
Yes
Natural
Non-sterile skirted and conical bottom tubes
Polypropylene
Screw Cap with O-ring
O-ring
Videos