Fisherbrand™ Colored Screw Caps

For use with nonsterile skirted- and conical-bottom tubes

Overview
Brand: Fisherbrand™

X500 SCREW CAP + O RING VIOLET

UNSPSC: 41120000

Code: A0

Additional Details:
Additional Details: Weight: 0.14000kg



Product Code. 11950089

Quantity Price
1 £ 34.85 / Pack of 500
Estimated Shipment
01-03-2017
Add to basket


Description and Specification

Specification

Autoclavable Yes
Product Type Screw Cap with O-ring
For Use With (Equipment) Non-sterile skirted and conical bottom tubes
Color Violet
Material Polypropylene
Includes O-ring