missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cheesecloth Wipers

Reagent-grade quality

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 0.22700kg

Product Code. 11704346

  • £87.45 / Pack of 120


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • 100% brilliant white, pure cotton construction will not scratch sensitive surfaces Have a high degree of cleanliness, softness, absorbency and strength
  • Can be used to safely clean microscope and camera optical parts, as well as delicate glassware, syringes, pipets, and pH electrodes
  • Absorb six times their weight in liquids and are useful for mopping up solvents and aqueous solutions
  • Inert to even harsh solvents; ideal as filtering material
  • Grade 10 (mesh size)

Order Info

Each wipe is 18 × 36 in. (46 × 91cm). Sixty wipes per dispenser box (two boxes per pack). Box dimensions: 9 1/2L × 9 1/2W × 4 1/2 in.H (24 × 24 × 11cm).

Compliance

Meet USP and Federal Specification CCC-G-101c.

Specifications

Specifications

USP, CCC-G-101c
18 x 36 in. (45.7 x 91.4cm)
Cotton
Microscopes, optical surfaces, Glassware, pH electrodes
White
45.7cm
Cheesecloth wipes
91.4cm
Videos
Documents