missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Phlebotomy Sample Collection Replacement Inserts

Brand:  Fisherbrand™

Code : 42

Additional Details : Weight : 0.97520kg

 View more versions of this product

Product Code. 11760634

  • £20.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Available for a 13 or 16mm tube rack
  • Insert is removable, washable, and disposable
  • Includes an extra-deep compartment for sharps container
  • Inserts sold in packs of four (4)
Specifications

Specifications

White
13mm tube rack and Blood Collection Tray (style A)
Polystyrene
Blood collection
Extra-deep compartment
Insert
Videos
Documentation