Learn More
Fisherbrand™ Phlebotomy Sample Collection Replacement Inserts
Brand:  Fisherbrand™
Code : 42
Additional Details : Weight : 0.94000kg
 View more versions of this product
Product Code. 11770634
  • £22.23 / Each
Estimated Shipment
06-07-2018
Add to basket
Description
Description
  • Available for a 13 or 16mm tube rack
  • Insert is removable, washable, and disposable
  • Includes an extra-deep compartment for sharps container
  • Inserts sold in packs of four (4)
Specifications
Specifications
White
16mm tube rack and Blood Collection Tray (style B)
Polystyrene
Blood collection
Extra-deep compartment
Insert
Videos
Documentation