missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Basic Protection Polypropylene Disposable Lab Coats with Elastic Cuffs

Latex-free spunbond polypropylene construction lab ware

Brand:  Fisherbrand™

21.14 GBP valid until 2018-08-31
Use promo code "13204" to get your promotional price.



Code : NEW

Additional Details : Weight : 3.40000kg

 View more versions of this product

Product Code. 15481922

  • £22.25 / Pack of 25


In Stock 3
Add to basket

Description

Description

  • Single-use lab coat
  • Standard collar, no pockets
  • Packed 25 lab coats/case
  • Elastic cuffs hold sleeves in place
  • Five snaps for a secure closure
  • Fabric weight: 50g/m2
  • White
Note: Other materials, colors and options are available. See Fisherbrand Basic Protection Disposable Lab Coats with Pockets, and Fisherbrand Basic Protection Disposable SMS Lab Coats in a variety of colors.

Specifications

Specifications

Standard
White
Polypropylene
3X-Large
Yes
Snap
Elastic
0
5-Snap
Videos
Documentation Special Offers

Special Offers

  • Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    : 13204

    2018-08-31

    *Discount will apply to qualifying orders received by Fisher Scientific between 04/06/2018 and 31/08/2018. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions