Learn More
Fisherbrand™ Autoclavable Bottletop Dispensers
Provides smooth, effortless dispensing
Brand: Fisherbrand™
Code : 41
Additional Details : Weight : 0.57600kg
Description
Provides smooth, effortless dispensing
- Wetted parts are PTFE, FEP and borosilicate glass (allows instrument to be used with a wide variety of chemicals)
- 7 volume ranges to choose from
- Bubble-free dispensing
- Effortless plunger movement
- Disassembles for easy cleaning and servicing
- Autoclavable at 121°C, 15 psi for 10 to 15 minutes
- Each bottle top dispenser includes one 28mm, 32mm, 36mm, 40mm, 45mm bottle neck adaptor
Component Description
- Piston: PTFE
- Cylinder: Glass
- Volume Adjustment Knob: Screw Type
- Valve Housing: PTFE
- Valve Assembly: Glass Ball & Seal
- Discharge Assembly: PTFE
- Delivery Tube: FEP
- Inlet Tube: FEP, 13 in. (with extension)
- Calibration: Individually calibrated and certified (in-lab by user is also possible)
- Accuracy & Reproducibility: In accordance with ISO 8655 standards
- Compability: Excellent with most reagents
Services
One year free replacement warranty against any manufacturing defects. The period of one year starts from the invoice date of Fisher Scientific to its customer.
Specifications
±0.6% (±0.015mL) | |
Autoclavable | |
PTFE, FEP, Borosilicate Glass | |
13-in. inlet tube (with extension) | |
±0.2% (±0.005mL) |
Analog Bottletop Dispenser | |
1 | |
0.25 to 2.5 mL | |
0.05 mL |