missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Amber Boston Round Bottles Without Cap

Ideal for UV sensitive contents. Fisherbrand™ Amber Boston Round Bottles without Cap are for general use.

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 20.45000kg

 View more versions of this product

Product Code. 11583632

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Features narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Amber color absorb all wavelengths in the ultraviolet spectrum
  • Ensure integrity for long term storage and protection for UV-sensitive contents
Specifications

Specifications

0.5 oz., 15mL
Amber
720/Cs.
No
18-400
Screw
15mL
Glass
Bottle, Boston Round
Narrow
Boston Round
Videos
Documentation