missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Amber Boston Round Bottles with White Polypropylene Pulp/Vinyl Lined Caps

Amber bottles are general use bottles ideal for when content integrity and UV protection is important. They ensure integrity for long term storage and offer excellent protection for UV-sensitive contents.

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 24.54000kg

 View more versions of this product

Product Code. 11553612

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

8 oz., 250mL
PTFE
Amber
108/PK.
White
Polyvinyl
Round
250mL
Poly/Vinyl
Glass
Yes
24-400
Narrow
screw cap
Videos
Documentation