missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Aluminum Weighing Dishes

Aluminum dishes for weighing and dispensing

Brand:  Fisherbrand™

Code : A0

Additional Details : Weight : 1.49400kg

 View more versions of this product

Product Code. 11789408

  • £250.26 / Pack of 1000
In Stock
to see stock.
Add to basket

Description

Description

Choose Fisherbrand™ Aluminum Weight Dishes for weighing and dispensing.

With Fluted sides

  • Also can serve as evaporating dishes or dust covers Lightweight, yet sturdy Convenient finger-grip handle can be etched prior to weighing for identification
With Smooth sides
  • Lightweight dishes with flanged rims Slightly sloping sides facilitate stacking
  • Especially useful for materials that cannot easily be cleaned from dish surface

Specifications

Specifications

Flat
Aluminum
Weighing Dish
13 mm
52 mm
20 mL
Fluted Sides
For weighing and dispensing
44 mm
44 mm (Inner Base), 52 mm (Inner Top)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers