Learn More
Fisherbrand™ Protective Disposable Face Masks
Provides convenient protection
£238.97 - £249.36
Specifications
Description Non-woven Polypropylene (outer and inner), Meltblown Polypropylene (filter)
Size Standard
Disposable Yes
Latex Free Yes
View More Specs
Products 4
Product Code Brand Color Includes Thickness Price Quantity & Availability  
Product Code Brand Color Includes Thickness Price Quantity & Availability  
15481932
View Documents
Fisherbrand™
Blue Tie closures 3-ply
£249.36
Pack of 500
View alternative products
Save Up to 
»
Estimated Shipment: 29-07-2021
Add to basket
15451932
View Documents
Fisherbrand™
White Ear loops 2-ply This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15471932
View Documents
Fisherbrand™
White Ear loops 3-ply This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15461932
View Documents
Fisherbrand™
Blue Ear loops 3-ply This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Provides convenient protection
  • Advanced protection
  • Disposable, single use
  • Choice of ear loops or tie closure
  • Nonwoven polypropylene outer layer
  • Thermally bonded nonwoven polypropylene inner layer
  • Latex-free
  • Choice of blue or white
  • Universal size/fit
Specifications
Specifications
Non-woven Polypropylene (outer and inner), Meltblown Polypropylene (filter)
Standard
Yes
Yes