missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Absorption Macro Rectangular Cells with PTFE Lid
Fit standard spectrophotometer cell holders
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 0.39330kg
Description
Fit standard spectrophotometer cell holders
- All cells are 12.5mm wide
- 1, 5 or 10mm path length rectangular cells
- Available with a variety of window materials for wide range of assays
- Available in four materials e.g. Quartz, Optical Quartz, Special Optical Glass and Optical Glass
- Quartz windows for transmission from far ultraviolet through the infrared
- Glass windows for routine work in ultraviolet region and visible region
Specifications
Macro Absorption Cell | |
Quartz | |
10mm | |
1 mm | |
52 mm | |
12.5 mm | |
Rectangle |
3.5 mL | |
PTFE | |
Ea., 8 Ea./Cs. | |
200 to 2500 nm | |
Standard Spectrophotometer Cell Holder | |
Transmission from far ultraviolet through the infrared |