Learn More
Fisherbrand™ 96-Well Polystyrene Plates
Ideal for antibiotic screens, cell-based assays and screening compounds.
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 10.93000kg
Description
Ideal for antibiotic screens, cell-based assays and screening compounds
Designed for compatibility with automated equipment
- Polystyrene
- Plates stack easily without lids
- Available in flat or round well shapes
- Nonsterile
- Untreated, provides uniform surface properties
- Use 12-565-511 lid (sold separately)
Specifications
Standard | |
White | |
400 μL | |
50-250 μL/well | |
Non-treated | |
Ideal for antibiotic screens, cell based assays and screening compounds |
8 x 12 | |
Polystyrene | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
180/Cs. |