missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ 96-Well Polypropylene Microplates
Ideal for storage—feature generous 500μL well volumes
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 3.07000kg
Description
- Polypropylene
- Minimum binding
- Resistant to many reagents
- Withstand temperatures from -80° to +121°C (-112° to +250°F)
- Automation-friendly design optimized for HTS
Specifications
Standard | |
Red | |
Polypropylene | |
96 | |
Round |
8 x 12 | |
96-well Storage Microplate | |
500 μL | |
Non-sterile | |
For storage feature generous 500μL well volumes |