Learn More
Fisherbrand™ 384-Well ShallowWell Polypropylene Microplates
Small volume applications
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 4.27000kg
Description
Feature a shared-wall design which yields increased well volume
- Automation compatible, ANSI standard
- Low-binding and chemically resistant surface
- Natural plates for storage or colorimetric assays
- White plates for luminescent assays, including SPA
- Black plates for fluorescent assays
- Round bottom wells for optimal sample recovery
- Rounded square wells minimize wicking (capillary action)
- Working volume range: 2 to 35μL/well
Order Info
Lids not included
Specifications
ANSI | |
16 x 24 | |
Polypropylene | |
384 | |
Round, Conical Bottom (V-well) |
Standard | |
White | |
120 μL | |
Non-sterile | |
Luminescent assays |