missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Horizontal Minigel Systems: Combs
For use with Horizontal Minigel systems
Brand: Fisherbrand™
Code : 04
Additional Details : Weight : 0.22000kg
Specifications
5 | |
Standard Comb | |
55μL |
1mm | |
Owl™ EasyCast™ B1A mini gel and A2-OK multi gel electrophoresis system | |
11.3mm |