missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for FB-VE16-1 Vertical Electrophoresis System

Brand:  Fisherbrand™

Code : 04

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11923035

  • £37.90 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

10
343μL
Comb
20cm
14cm
13cm
0.15cm
Owl™ P1-CST multiple gradient gel caster
18cm
16cm
1.04cm
16 x 14cm
Videos