missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Bottles

Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer

Brand:  Thermo Scientific™ N10805

Code : X3

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11747996

  • £253.00 / Each
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer

For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer

  • Subsitute bottles of different sizes for added convenience
  • Wash and waste bottles include liquid level sensors
  • 4L wash and 9L waste bottles available for automation applications

ELISA-based assays, cell washing

Specifications

Specifications

2 L
67.62 oz.
Wellwash Microplate Strip Washer
Wash Bottle
Cap with tubing and liquid level sensors
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers