Learn More
Fisherbrand™ Traceable™ International Standards Extreme-Accuracy Digital Thermometer
Traceable™ International Standards Extreme-Accuracy Digital Thermometers ideal for measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods, and wastewater.
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 0.20000kg
Description
Traceable™ International Standards Extreme-Accuracy Digital Thermometers ideal for measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods, and wastewater.
Units read 0.00, 25.00, 37.00, or a combination of all three in °F and °C.
- Provides accurate readings with tip penetration of 0.33 in.
- Records time/date for min/max readings and hi/lo alarm alerts
- Displays minimum/maximum and current temperatures
- Min/max monitors readings over any period; displays exact time/date of min/max temperature
- Alarm provides two visual LEDs and two audio alerts when temperature exceeds set points; displays exact time/date of dual thermometer alarms
- Programmable in 0.1° increments; alarms signal continuously even if temperature returns to non-alarm range
- Resolution: 0.01°
- Accuracy: ±0.05°C within ±2°C of standard
- Probe Dia.: 0.12 in. (0.30 cm)
- Stem Length: 6.25 in. (15.87 cm)
- Overall Length: 9 in. (22.86 cm)
- Cable: 5 ft. (1.5 m)
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.
Compliance
Meets ITS-90, International Temperature Standard.
Specifications
ISO 17025, A2LA, NIST | |
4.25 x 2.75 x 0.75 in. (10.7 x 6.9 x 1.9 cm) | |
0.75 in. | |
Stainless Steel probe, High-impact, chemical-resistant ABS plastic case, stand, Velcro™, magnetic strips, wall mount, two AA batteries (allow continuous, 1-year always-on monitoring) | |
107 mm | |
ABS Plastic | |
5 oz. | |
2.75 in. | |
80.6°F | |
73.4°F | |
73.4°F to 80.6°F | |
±0.05°C | |
6.9 cm |
0.12 in. | |
Measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods and wastewater | |
1.9 cm | |
6.25 in. | |
15.9 cm | |
0.01°C | |
140 g | |
4.25 in. | |
27°C | |
23°C | |
23.00°C to 27.00°C | |
LED |