Learn More
Fisherbrand™ Test Tube Holders for Heavy-duty Vortex Mixer
For use with heavy-duty vortex mixer.
Brand: Fisherbrand™
Additional Details : Weight : 0.08000kg
Description
- Holds a wide range of test tube numbers and diameters
- Requires Universal Holder
Specifications
Test Tube Holder | |
Heavy-duty vortex mixer | |
20 |
15 to 18 mm | |
Foam insert | |
Universal holder |