Learn More
Fisherbrand™ Standard Exhaust Oil Filters
As supplied with Fisher Scientific Maxima C Plus pumps
Brand:  Fisherbrand™
Code : 45
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11847044
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
As supplied with Fisher Scientific Maxima C Plus pumps
These compact, round filters simply screw into threaded exhaust port to collect oil expelled during operation. A replaceable filter element within the oil filter separates the oil into particles no larger than 0.3μm, and drops collected oil back into the oil case.
  • Filter element is easily replaced
  • Reorder number located on filter canister
1For M4C or M8C vacuum pumps designed before September 1999 (S/N before FKK090000).
2For M4C or M8C vacuum pumps designed after August 1999 (S/N FKK090000 and newer).
Specifications
Specifications
2-1/2 in.
Model M2C and M4C vacuum pump
6.35 cm
63.5 mm
2.5 in.
Standard Exhaust Oil Filter