missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Spider Support Stands for Overhead Stirrers

Able to clamp on the edge of pails, drums, square or rectangular tanks.

Brand:  Fisherbrand™

Code : 04

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15162997

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Provides a safe and secure mounting system on any vessel

  • Fits securely on vessels with rims up to 5/8 in. wide
  • Adjustable legs and swivel clamp
  • Light weight aluminum base with a 5/8 in. diameter and 24 in. high stainless steel support rod

Specifications

Specifications

0.62 in.
Vessel up to 15 in. inscribed circle
71.2 cm
1.58 cm
28 in.
Spider Support Stand
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers