Learn More
Fisherbrand™ Single-Action Stopwatch™
Simple and intuitive to use.
Brand:  Fisherbrand™
Code : 50
Additional Details : Weight : 0.12600kg
Product Code. 11547463
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Start, stop and reset are all achieved with a single button
  • Provides single-action timing up to 15 minutes with 0.1-second resolution
  • Extra-tough metal housing
Specifications
Specifications
Single-action Stopwatch
0.1 sec.
3 oz.
Lab and plant use, provides single-action timing up to +15 min. with 0.1 sec. resolution
15 min.
5.08 x 1.27 cm
85.04 g