missing translation for 'onlineSavingsMsg'
Learn More

Bibby Scientific Microplate 96 well PTFE 9 x 8mm I.D. 1.1mL per well

Brand:  Bibby Scientific PS80048

Microplate 96 well PTFE 9 x 8mm I.D. 1.1mL per well

Code : 04

Additional Details : Weight : 1.00000kg

Product Code. 12719038

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.