missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ MultiTube Vortexers: Centrifuge Tube Racks

Centrifuge tube racks for Fisherbrand™ MultiTube Vortexers.

Brand:  Fisherbrand™

Additional Details : Weight : 0.04500kg

 View more versions of this product

Product Code. 15519014

  • £53.80 / Each
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

Centrifuge tube racks for Fisherbrand™ MultiTube Vortexers.

For use with Fisher Scientific MultiTube Vortexers

Color-coded foam racks for centrifuge tubes

Specifications

Specifications

50 mL
Centrifuge Tube Rack
Multitube vortexer
50 mL
Red
17
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers