Learn More
Fisher Scientific™ Maxima C Plus Gasket Sets
For use with Model M16C vacuum pump.
Brand:  Fisher Scientific™ 01-257-3C
UNSPSC : 41105101
Code : 45
Additional Details : Weight : 0.01000kg
Product Code. 12346657
  • £256.50 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications
Description
For use with Model M16C vacuum pump
Description & Specifications
Specifications
Gasket Set
Model M16C vacuum pump
Viton™ O-rings, shaft seals and gaskets