Fisher Scientific™ Key Chain Laser Pointers

For slide presentations, microscope projections, field studies, wall charts and lectures

Overview
Brand: Fisher Scientific™

Manufacturer Part Number: 3141

UNSPSC: 41115307

Code: 46

Additional Details:
Additional Details: Weight: 1.00000kg



Product Code. 11784366

Quantity Price
1 £ 66.25 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket


Description and Specification

Key Chain Micro Laser Pointer

  • Half the size of pocket laser
  • Features output power of 4mW and range of 300 ft. (91m)
  • Brilliant red beam with 3.5mm diameter can be seen under normal lighting conditions
  • 2.37L × 0.56 in. dia. (6.0 x 1.4cm) body can be carried securely on keychain
  • Switch is momentary ON/OFF
Laser Key Chain
  • Brilliant red beam works equally well in daylight and dark
  • Easy to carry with attached key chain
  • Highly visible 650nm wavelength
  • Diameter, 3.5mm
  • Output, 4 mW Range, 400 ft.
  • Dimensions, 2.5 × 1 × 0.25 in. (6.35 × 2.54 × 0.64cm)
  • Weight, 0.5 oz. (14.2g)

Documentation

Technical documents