Learn More
Fisherbrand™ Isotemp™ Shake Touch Shakers Accessory Blocks
Interchangeable blocks to accommodate a wide variety of tubes and microplates for use with Fisherbrand™ Isotemp™ Shake Touch shakers.
Brand: Fisherbrand™
Additional Details : Weight : 0.50000kg
Description
Interchangeable blocks to accommodate a wide variety of tubes and microplates for use with Fisherbrand™ Isotemp™ Shake Touch shakers.
- Well plate blocks with lid
- PCR plate blocks with lid
- Tube blocks
- Conical tube blocks
- Microplate thermal blocks
Specifications
15 mL | |
4.02 in. | |
0.7 in. | |
4.2 x 5.8 x 5.0 in. (10.7 x 14.7 x 12.7 cm | |
Heating/shaking | |
9 |
Shaker Accessory Block | |
10.2 cm | |
1.7 cm | |
Conical tube block for 15 mL tubes | |
Shake Touch Shakers |