missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Isotemp™ Shake Touch Shakers Accessory Blocks

Interchangeable blocks to accommodate a wide variety of tubes and microplates for use with Fisherbrand™ Isotemp™ Shake Touch shakers.

Brand:  Fisherbrand™

Additional Details : Weight : 0.40000kg

 View more versions of this product

Product Code. 15569044

  • £470.00 / Each


Estimated Shipment
31-03-2020
Add to basket

Description

Description

Accessories Offered:
  • Well plate blocks with lid
  • PCR plate blocks with lid
  • Tube blocks
  • Conical tube blocks
  • Microplate thermal blocks
Specifications

Specifications

24-well tube block for 1.5mL tubes
Includes rack and cover
Shake Touch Shakers
1.1cm
Heating/shaking
3.35cm
1.5mL
4.0 x 5.6 x 2.1” (10.2 x 14.2 x 5.3cm)
1.5mL
24
Videos
Documents