Learn More
Fisherbrand™ Isotemp™ Shake Touch Shakers Accessory Blocks
Interchangeable blocks to accommodate a wide variety of tubes and microplates for use with Fisherbrand™ Isotemp™ Shake Touch shakers.
Brand: Fisherbrand™
Additional Details : Weight : 0.40000kg
Description
Interchangeable blocks to accommodate a wide variety of tubes and microplates for use with Fisherbrand™ Isotemp™ Shake Touch shakers.
- Well plate blocks with lid
- PCR plate blocks with lid
- Tube blocks
- Conical tube blocks
- Microplate thermal blocks
Specifications
0.5 mL | |
2.57 cm | |
0.8 cm | |
Tube block for 0.5 mL tubes | |
Shake Touch Shakers | |
30 |
1.01 in. | |
0.312 in. | |
4.0 x 5.6 x 1.8 in. (10.2 x 14.2 x 4.6 cm) | |
Heating/shaking | |
Rack and cover |