Learn More
Fisherbrand™ Isotemp™ Hotplate
Experience exceptional safety and temperature performance for your routine protocols.
£174.00 - £311.00
Specifications
Description Hotplate
Hertz 50/60Hz
View More Specs
Products 8
Product Code Brand Top Plate Material Electrical Requirements Plug Type Price Quantity & Availability  
Product Code Brand Top Plate Material Electrical Requirements Plug Type Price Quantity & Availability  
15313518 View accessory products Fisherbrand™
Ceramic 230V 50/60Hz EU £192.00
Each
Estimated Shipment
31-01-2019
Add to basket
View alternative products »
15313538 View accessory products Fisherbrand™
Ceramic 230V 50/60Hz EU £303.00
Each
In Stock 1
Add to basket
15323518 View accessory products Fisherbrand™
Ceramic 230V 50/60Hz UK/AUS/CN £201.00
Each
In Stock 16
Add to basket
View alternative products »
15323538 View accessory products Fisherbrand™
Ceramic 230V 50/60Hz UK/AUS/CN £311.00
Each
In Stock 6
Add to basket
15343508 View accessory products Fisherbrand™
Ceramic 230V 50/60Hz EU £174.00
Each
Estimated Shipment
31-01-2019
Add to basket
View alternative products »
15353508 View accessory products Fisherbrand™
Ceramic 230V 50/60Hz UK/AUS/CN £182.00
Each
In Stock 1
Add to basket
View alternative products »
15373518 View accessory products Fisherbrand™
Aluminum 230V 50/60Hz EU £239.00
Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
View alternative products »
15383518 View accessory products Fisherbrand™
Aluminum 230V 50/60Hz UK/AUS/CN £247.00
Each
In Stock 4
Add to basket
View alternative products »
Description
Description
Advanced technology
  • Strong magnetic coupling
  • Temperature adjustable in 1 degree increments
  • HOT TOP warning system designed to protect from accidental burns with prominent display when heating surface is above 50°C (122°F)
  • Automatic over-temperature protection
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Raised display design protects electronics from spills
  • Optional temperature probe and splash guard protection shield
  • Variable sizes for different sample volumes
Certifications
cCSAus (100-120V models), CE (230V models)
Compliance
RoHS compliant
Warranty
Three years
Specifications
Specifications
Hotplate
50/60Hz