Learn More
Fisherbrand™ Isotemp™ Stirring Hotplate
Experience precise stirring control, exceptional safety and temperature performance for your routine protocols.
£290.00 - £392.00
Specifications
Description Hotplate Stirrer
View More Specs
Products 4
Product Code Brand Top Plate Material Plug Type Max. Temperature (Metric) Surface Area (Metric) Heating Price Quantity & Availability  
Product Code Brand Top Plate Material Plug Type Max. Temperature (Metric) Surface Area (Metric) Heating Price Quantity & Availability  
15303518 View accessory products View Documents Fisherbrand™
Ceramic UK/AUS/CN 540°C 10.8 x 10.8cm £290.00
Each View alternative products
Save Up to 
»
In Stock 9
15363518 View accessory products View Documents Fisherbrand™
Ceramic UK/AUS/CN 540°C 18.4 x 18.4cm £325.00
Each View alternative products
Save Up to 
»
In Stock 8
15363538 View accessory products View Documents Fisherbrand™
Ceramic UK/AUS/CN 400°C 26.0 x 26.0cm £392.00
Each View alternative products
Save Up to 
»
In Stock 3
15343528 View accessory products View Documents Fisherbrand™
Aluminum UK/AUS/CN 300°C 18.4 x 18.4cm £366.00
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Description
Description
Advanced technology
  • Slow-speed stirring, consistent speed control, and strong magnetic coupling
  • Temperature adjustable in 1 degree increments
  • HOT TOP warning system designed to protect from accidental burns with prominent display when heating surface is above 50°C (122°F)
  • Automatic over-temperature protection
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Raised display design protects electronics from spills
  • Optional temperature probe and splash guard protection shield
  • Variable sizes for different sample volumes
  • Includes stir bar
Certifications
cCSAus (100-120V models), CE (230V models)
Compliance
RoHS compliant
Warranty
Three years
Specifications
Specifications
Hotplate Stirrer