missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Isotemp™ Heating Blocks

For Isotemp digital models 125DQ, 145DQ and analog dry-bath incubators

Brand:  Fisherbrand™

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11880740

  • £84.71 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Choose from blocks that hold 6 to 25mm O.D. standard tubes, 15mL or 50mL conical-bottom tubes, microcentrifuge tubes.

  • Solid aluminum construction
  • Sample wells are shaped so that heat is evenly transmitted to all parts of the sample tube
  • Shape also eliminates the temperature gradient that may invalidate tests

Specifications

Specifications

Isotemp 125DQ, 145DQ and economical dry-bath incubator
5.1cm
Aluminum
9.5cm
For 8 x 20mm tube
51mm
2.9 x 3.75 x 2 in. (7.4 x 9.5 x 5.1cm)
7.4cm
5cm
7.4cm
9.5cm
50mm
Heating Block
74mm
7.4 x 9.5 x 5cm
Videos