Learn More
Fisherbrand™ Elite™ Multichannel Pipetters
New, improved ergonomic design provides newly designed finger rest and index finger and hand support for maximum comfort
£493.22 - £560.43
Specifications
Warranty 3 years
View More Specs
Products 4
Product Code Brand Volume (Metric) Number of Channels Price Quantity & Availability  
Product Code Brand Volume (Metric) Number of Channels Price Quantity & Availability  
11835772
View accessory products View Documents
Fisherbrand™
5 to 50 μL 8
£493.22
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-09-2021
Add to basket
11845772
View accessory products View Documents
Fisherbrand™
30 to 300 μL 8
£493.22
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-08-2021
Add to basket
11815772
View accessory products View Documents
Fisherbrand™
1 to 10 μL 8
£493.22
Each
View alternative products
Save Up to 
»
Estimated Shipment: 22-07-2021
Add to basket
11885772
View accessory products View Documents
Fisherbrand™
30 to 300 μL 12
£560.43
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-09-2021
Add to basket
Description
Description
  • Fully autoclavable - entire pipetter can be steam autoclaved
  • Soft-touch tip ejection—a gearing mechanism reducing tip ejection force up to 50%
  • Ergonomic comfortable lightweight handle - comfortable grip and effortless tip attachment
  • Optimal fit with SureOne pipetter tips - best pipetting performance
  • Super blow-out piston - creates 150% bigger air push than conventional piston for delivery of the micro size drops
  • 8- and 12-channel pipetters with volume range of 1 up to 300μL
  • Extremely low plunger forces - work longer without fatigue and reduce the risk of RSI
Specifications
Specifications
3 years