Learn More
Fisherbrand™ Traceable™ Digital Thermometers with Stainless-Steel Stem, 0.25 in. LCD Screen, and Protective Guard
Easy-to-read handheld digital thermometers
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 0.05220kg
Description
Fisherbrand™ Traceable™ Digital Thermometers with Stainless-Steel Stem are resistant to acids, bases, solvents, and may be used to measure temperature of gases, food, and other semisolids.
- Longer probes are excellent for measuring sample temperatures while stirring in deep vessels or water baths
- Thermometers with longer sampling rates allow thermometer to be removed from sample before reading—ideal when display is difficult to read in measuring position
- For use in cuvets, soil, incubators, petroleum products and foods
Certifications
Multi-point calibration on individually-numbered Traceable Certificate assures accuracy from ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA™; indicates traceability to NIST™.
Specifications
0.14 in. | |
Traceable certificate, Stainless steel probe | |
365 mm | |
ABS Plastic | |
1 oz. | |
14.38 in. | |
300°C | |
-50°C | |
-50°C to +300°C | |
Digital LCD |
Read temperatures in foods, gases, samples, reagents, water baths and semisolids | |
11.37 in. | |
28.87 cm | |
0.1°C | |
20 g | |
572°F | |
-58°F | |
-58°F to +572°F | |
±0.5°C |