Learn More
Fisherbrand™ Air-Powered Analog Overhead Stirrers
Variable speed and explosion-proof stirrers ideal for safely mixing all types of solvents, lacquers and volatile chemicals without the possiblity of spark ignition.
Brand: Fisherbrand™
Code : 04
Additional Details : Weight : 2.70000kg
Description
- Needle valve allows variation in mixing speed
- Special muffler ensures quieter operation
- Reversible models have motors which can be set to rotate in either direction by connecting the air supply line to the right port for counter-clockwise rotation or the left port for clockwise rotation
- Can be used with flammable chemicals for explosion proof mixing
Specifications
1200 rpm | |
0.37 in. | |
0.3 | |
10 L x 0.56 in. dia. support rod, 4 ft. air hose with double fittings for connection to air supply | |
203 | |
6 lb. |
20 L | |
0.95 cm | |
Safely mixing all types of solvents, lacquers and volatile chemicals | |
30 to 80 psi | |
10,000 mPas | |
2.7 kg |