Learn More
Fisherbrand™ accumet™ XL250 Benchtop pH/ISE Dual Channel Meters
Provide advanced pH and ion measurements with simple operation and offer a large, vivid color touch-screen display. Fisherbrand accumet XL250 pH/ISE dual channel benchtop meters offer advanced features with simple operation to help ensure your measurements are reliable and your laboratory processes are efficient.
Brand: Fisherbrand™
Code : 43
Additional Details : Weight : 0.10000kg
Includes: accumet XL250 pH/ISE dual channel benchtop meter, ammonia electrode (13-620-509), TRIS-compatible double junction refillable glass-body pH electrode (13-620-183A), ATC probe (13-620-19), electrode stand, RS232 and USB cables, 100/240V universal power supply
Description
Provide advanced pH and ion measurements with simple operation and offer a large, vivid color touch-screen display. Fisherbrand accumet XL250 pH/ISE dual channel benchtop meters offer advanced features with simple operation to help ensure your measurements are reliable and your laboratory processes are efficient.
- High resolution color display shows data clearly from wide viewing angles and distances
- Optimized touch-screen with glass/glass sensitivity provides great ease of use
- Onscreen info includes date and time, stability indicator, alarms and context-specific “help” screens
- One to six point pH calibration with 0.001 pH resolution
- Automatic buffer recognition with six pH buffer sets or custom buffer option
- Two to six point ISE calibration with 0.001 ppm, mg/L or molar resolution
- ISE methods for known addition, known subtraction, analyte addition and analyte subtraction
- Automatic or manual temperature compensation, displayed in °C or °F
- Display two pH or ISE measurements simultaneously with dual channel BNC inputs
- Up to 10 password protected user ID accounts, each retains unique calibration and instrument settings with memory to store 2000 data points
- RS232 and USB interfaces for printer or computer interfacing and real-time onscreen graphing of measurements
- RJ45 Ethernet/Internet port to connect Local Area Network (LAN) or Internet
- Bidirectional communication for LIMS connectivity and 3rd party software
- Connectivity for peripherals such as keyboard, mouse or other USB device
- Multilanguage interface with English, German, Chinese, French, Italian and Spanish
- Two ports for optional stirring probe (Cat. No. 13-620-BSP) with five speed settings
- Option to wall-mount meter to create additional bench space
- Help meet ISO, FDA, USDA and EPA quality assurance guidelines
- Includes meter accuracy at five test points across pH range
- Includes meter accuracy at five test points across corresponding mV range
- Testing performed by accredited laboratory, includes NIST-Traceable certificate
- pH Range: -2.000 to 20.000 pH
- pH Resolution: 0.1, 0.01, 0.001 pH
- pH Accuracy: ±0.002 pH
- ISE Range: 0.001 to 19999 ppm, mg/L or molar
- ISE Resolution: 0.001, 0.01, 0.1, 1 (automatic)
- ISE Accuracy: 0.5 % full scale (monovalent ion); 1 % full scale (divalent ion)
- mV Range: ±2000.0mV
- mV Resolution: 0.1mV
- mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
- mV Offset Adjustment: Up to ±150mV
- Temperature Range: -5° to +105°C (23° to 221.0°F)
- Temperature Resolution: 0.1°C (0.1°F)
- Temperature Accuracy: ±0.2°C (±0.3°F)
- Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
- ISE Calibration Points: 2 to 6 points
- ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
- Memory: 2000 data sets per user ID with date/time stamp
- Self-Test: Yes
- Inputs: Two BNC, 2 mini-phone ATC, ground
- Power Requirements: 100 to 240VAC, 50/60Hz
- Dimensions (L x W x H): 24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.)
- Weight: 1.13kg (2.5 lb.)
Certifications
CE, TUV 3-1, FCC Class A
Services
3 years on meter
Specifications
RS232, USB-A, Mini-B USB, RJ45, stirrer port (2) | |
0.5% full scale (monovalent ion), 1% full scale (divalent ion) | |
±0.002 | |
Up to 6 preset or 5 custom | |
3 to 86400 sec. | |
Multiple | |
LCD | |
±2000 mV | |
2/3/4 Digits | |
-5°C to +105°C | |
2 BNC, 2 ATC | |
±2000 mV | |
0.1mV | |
0.1°C | |
24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.) | |
24.6 cm | |
3.89 in. | |
2.5 lb. | |
100/240V |
Multiple | |
±0.2 mV | |
2 to 6 | |
NIST, ISO, FDA, USDA, EPA | |
2000 | |
2 | |
pH, mV, ISE, Ion, Concentration, Temperature | |
-2 to 20 | |
0.1/0.01/0.001 | |
Meter, 13-620-509 ammonia electrode, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), electrode arm, RS-232 and USB cables, and 110/220V power supply | |
XL250 Dual pH/ISE Ammonia Kit | |
Multiple | |
±0.3°C | |
3 Years | |
9.68 in. | |
17.3 cm | |
9.9 cm | |
1.13 kg | |
50/60Hz |