Learn More
Fisherbrand™ accumet™ Temperature-Compensated Two-Cell Conductivity Probes - Mercury - Free
For use on accumet™ Excel (XL) series benchtop meters
Brand:  Fisherbrand™
Code : 43
Additional Details : Weight : 0.10000kg
Product Code. 11540164
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Includes: 8-pin connectors
Description
Description
For use on accumet™ Excel (XL) series benchtop meters
  • Immersion-type cell
  • Use in standard electrode holder: unique compact size is same as typical pH electrode
  • Built-in temperature sensors provide temperature-compensated readings
  • Epoxy body