Fisher Scientific™ accumet™ Solid-State Half-Cell ISEs - Mercury - Free

For bromide, chloride, fluoride, lead and silver/sulfide determinations

Overview
Brand: Fisher Scientific™

Manufacturer Part Number: 93X561101

SOLID-STATE HALF-CELL ISE SILVER/SULPHide determination epoxy body BNC connector 13mm

UNSPSC: 41115608

Code: 43

Additional Details:
Additional Details: Weight: 0.10000kg



Product Code. 11550144

Quantity Price
1 £ 509.47 / Each
Estimated Shipment
14-03-2017
Add to basket


Description and Specification

Specification

Connector Types BNC
Applications Determination of Silver/Sulfide concentration in aqueous solutions, certain nonaqueous solutions, water, pulping liquors
Material Epoxy
pH Range 2 to 12