missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ Micro and Semimicro Size Glass Combination Electrodes - Mercury - Free
Ideal for microwell plates, serum cups, cuvets, micro test tubes, NMR tubes and capillary tubes
£247.00 - £278.00
Specifications
Junction | Single |
---|---|
Material | Glass |
pH Range | 0 to 14 |
Temperature (Metric) Operating | -5°C to +110°C |
Filling Solution Type | Saturated KCl with AgCl. KCl |
Product Code | Brand | Connector Types | Length (Metric) | Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Connector Types | Length (Metric) | Type | Price | Quantity & Availability | |||
11590104
|
Fisherbrand™
|
BNC | 100 mm | Combination pH electrode, Ag/AgCl |
£247.00
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||
11500114
|
Fisherbrand™
|
BNC | 150 mm | pH/ORP Combination Titration Electrode, Extra-Long Ag/AgCl |
£278.00
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||
11540064
|
Fisherbrand™
|
U.S. Standard | 100 mm | pH/ORP Combination Titration Electrode, Standard Ag/AgCl |
£273.00
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||
11550064
|
Fisherbrand™
|
U.S. Standard | 150 mm | pH/ORP Combination Titration Electrode, Extra-Long Ag/AgCl |
£269.00
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||
11560064
|
Fisherbrand™
|
U.S. Standard | 60 mm | pH/ORP Combination Titration Electrode, Miniature Ag/AgCl |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Ideal for microwell plates, serum cups, cuvets, micro test tubes, NMR tubes and capillary tubes
Zero to 14pH rangeSpecifications
Single | |
0 to 14 | |
Saturated KCl with AgCl. KCl | |
Ag/AgCl | |
-5°C to +110°C | |
1 Year |
Glass | |
-5°C to +110°C | |
Small samples | |
23°F to 230°F | |
Micro |