missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ Glass Body Standard Size Combination Electrodes - Mercury-Free
For routine pH and titration measurements with small samples and small vessels, where separate half-cell electrodes may be impractical
£169.00 - £227.00
Specifications
Closure Type | Twist Open/Twist Close |
---|---|
Material | Glass |
pH Range | 0 to 14 |
Type | pH Combination Electrode |
Internal Reference | Ag/AgCl |
Product Code | Brand | Connector Types | Junction | Filling Solution Type | Length (Metric) Cable | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Connector Types | Junction | Filling Solution Type | Length (Metric) Cable | Price | Quantity & Availability | ||
10232781
|
Fisherbrand™
|
BNC | Single Ceramic | SP135-500 | 76.2 cm |
£169.00
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||
11560174
|
Fisherbrand™
|
BNC | Double Ceramic | SP138-500 | 76.2 cm |
£201.00
Each |
Estimated Shipment: 08-05-2024
Log in to see stock. |
||
11520064
|
Fisherbrand™
|
U.S. Standard/Pin | Single Ceramic | SP135-500 | 76.2 cm |
£227.00
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||
11530064
|
Fisherbrand™
|
U.S. Standard/Pin | Single Ceramic | SP135-500 | 152.4 cm |
£221.00
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||
Description
For routine pH and titration measurements with small samples and small vessels, where separate half-cell electrodes may be impractical
- 0 to 14pH range
- Porous ceramic junction
- Twist-open/twist-close cap for easy filling
- Constant distance between FS-5 Universal Glass pH bulb and reference junction means high reproducibility
- Immersion depth, ∽4 in. (10.1cm)
- 30 in. long (76.2cm) leads, except 13-620-91, which has 60 in. long (152.4cm) lead
Choice of Ag/AgCl Single- or Double-Junction internal reference elements
- Single-junction models with Ag/AgCl reference are perfect for routine pH and titration measurements of aqueous solutions with small samples, small vessels
- Double-junction models with Ag/AgCl reference are useful in aqueous and nonaqueous solutions, and in applications requiring TRIS buffers
- For aqueous and nonaqueous samples
- Double-junction Ag/AgCl eliminate measurement and clogging problems when used in research with Tris buffers
Specifications
Twist Open/Twist Close | |
0 to 14 | |
Ag/AgCl | |
4 in. |
Glass | |
pH Combination Electrode | |
-5°C to +100°C | |
101.6 mm |