missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ Gel-Filled Paper Pulp Double-Junction pH Combination Electrodes: Mercury-Free
Dual junctions provide fast response and long life in paper pulp testing
£190.00 - £201.00
Specifications
Junction | Dual |
---|---|
Material | Gel-filled Polymer/Polypropylene |
pH Range | 0 to 14 |
Type | pH Combination Electrode |
Filling Solution Type | Gel |
Product Code | Brand | Connector Types | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Connector Types | Price | Quantity & Availability | |||||
11530194
|
Fisherbrand™
|
BNC |
£190.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11590154
|
Fisherbrand™
|
U.S. Standard/Pin |
£201.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
Description
Dual junctions provide fast response and long life in paper pulp testing
- Perfect for paper pulp testing, mine assay labs, smelters—anywhere junction clogging may be a problem
- Inner chamber houses reference element with gelled KCl and AgCl; the outer chamber contains KCl only
- Eliminates heavy metal ions that react with sulfide ions in samples such as black and green liquors
- No insoluble sulfide precipitates to block electrode junction and cause premature failure
- Polypropylene body
- Fisher Scientific universal glass membrane for rapid, reproducible response
- Glass tip is recessed in end
- Accuracy, ±0.05pH at 25°C (77°F) Efficiency, >95% Sodium Ion error, <±0.05pH at 25°C (77°F) Response time, 30 seconds Zero potential, 7.0 ±0.3pH
- Membrane resistance, <100megohms at 25°C (77°F)
- Ideal for routine process monitoring in paper pulp plants, mine assay labs, smelters
- Fits any application where the presence of S= in samples causes problems of slow response and junction failure
Specifications
Dual | |
0 to 14 | |
Gel | |
pH meter | |
30 sec. |
Gel-filled Polymer/Polypropylene | |
pH Combination Electrode | |
Ideal for routine process monitoring in paper pulp plants, mine assay labs, smelters | |
±0.05 |