Learn More
Fisherbrand™ accumet™ BOD Probe for accumet Benchtop Dissolved Oxygen Meters, Mercury-Free
Self-stirring BOD Probe for all current accumet benchtop meters with dissolved oxygen mode.
Brand: Fisherbrand™
Code : 43
Additional Details : Weight : 0.10000kg
Description
Self-stirring BOD Probe for all current accumet bench-top meters with dissolved oxygen mode. For use with Accumet bench-top and portable meter
- Probe type: BOD
Specifications
Accumet Benchtop and Portable Meter | |
BOD, DO | |
BOD |