Learn More
Fisherbrand™ accumet™ AB15 Basic and BioBasic™ pH/mV/°C Meters
Simple, menu-guided operation in an affordable pH/mV meter
Brand: Fisherbrand™
Code : 43
Additional Details : Weight : 0.10000kg
Includes: Meter Only package includes meter, power supply, electrode support (13-637-671) and operation manual.
Meter Kit package includes items above plus an accumet liquid-filled, glass body combination pH Ag/AgCl Electrode (13-620-285) and separate ATC probe (13-620-19).
BioBasic Meter Kit package for the life science lab includes meter, power supply, manual, electrode support and accuTupH™ rugged bulb, glass body combination pH Double-Junction Electrode (13-620-183) and separate ATC probe (13-620-19).
University Meter Kit package includes meter; pH/ATC plastic body refillable electrode (13-620-631), electrode support (13-637-671), operation manual and power supply.
Some packages are available with a Certificate of Traceability to NIST™ Standards.
Description
Easy operation, accuracy and affordability
- Large LCD guides user with plain-language prompts and error messages
- Display indicators keep you apprised of menu choice, measuring units, buffers standardized (in pH mode) and temperature
- No cryptic icons to learn: prompts and status/error messages are displayed in plain language
- Store up to 50 datapoints in nonvolatile memory
- Powerful pH mode offers selectable resolution to 0.01pH
- Automatic recognition of 15 buffers and five-point calibration
- BNC and reference pin electrode inputs, ATC probe input, and input for accuFet™ electrode
Order Info
Dimensions: 7.5L x 5.5W x 3.25 in.H (19 x 14 x 8cm). Net weight: 1.875 lb. (0.8kg).
Service Plan
Service agreement available.
Services
accumet Model AB15 is warranted to the end user to be free from defects in materials and workmanship under normal use for a period of three years.
Specifications
pH/ATC Electrode 13-620-631, electrode support arm and bracket; manual | |
LCD | |
Multiple | |
120V, 50/60Hz | |
60 Hz |
pH, mV, Temperature | |
Multiple | |
Multiple | |
120 V |