missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ YPD Broth (Powder/Molecular Genetics)
Brand: Fisher BioReagents™ BP2469-10
Code : A0
Additional Details : Weight : 10.05000kg
Specifications
4°C | |
Molecular genetics | |
Microbiology Media | |
10,000 g |
Powder | |
YPD Broth | |
Poly Bottle |