Learn More
Fisher BioReagents™ OptiGrow™ Preweighed Media
Optimized for superior yield of bacterial culture
Brand:  Fisher BioReagents™ BP1426-10P1
Code : 32
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 11820715
  • £80.96 / Each
Estimated Shipment
27-07-2018
Add to basket
Description
Description
Four classic media, in convenient, preweighed pouches.
  • Preblended growth-promoting components save time and preparation costs
  • Reduce formulation errors
  • Each pouch contains enough material for 1 liter of liquid media
  • No more weighing of dusty powders
  • High quality—assured by stringent testing procedures
  • LB is used for the culturing of E. coli in molecular biology procedures
  • Terrific Broth is a buffered enriched medium used to obtain high yields of plasmid DNA from transformed E. coli
Order Info
Supplied as 10 pouches per box.
Specifications
Specifications
Molecular genetics
LB Broth, Miller
10 Pouches
LB broth, Miller
Solid
Pouch
4°C
Culturing of Escherichia coli
Videos
SDS Documentation