missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ NZY Broth (Powder/Molecular Genetics)

Brand:  Fisherbrand™

Code : 32

Additional Details : Weight : 0.55000kg

 View more versions of this product

Product Code. 10326603

  • £104.10 / 500g


Estimated Shipment
12-02-2018
Add to basket

Specifications

Specifications

Molecular genetics
NZY Broth
500g
NZY Broth
Powder
Poly Bottle
4°C
Videos
SDS Documentation