missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Molecular Weight Markers
Suitable for determining the size of DNA fragments
£91.94 - £196.00
Specifications
Color | Colorless |
---|---|
Product Type | DNA Molecular Weight Marker/Ladder |
For Use With (Application) | Determining the size of DNA fragment |
Product Code | Brand | Description | Form | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Form | Quantity | Price | Quantity & Availability | |||
10428543
|
Fisher BioReagents™
BP2551-50 |
100 bp DNA Ladder | Liquid | 50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11483084
|
Fisher BioReagents™
BP2554-25 |
lambda DNA/Hind III/PhiX-174 RF DNA-Hae III Digest | Lyophilized | 25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10523325
|
Fisher BioReagents™
BP2553-100 |
Kilobase DNA Marker | Liquid | 50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11468473
|
Fisher BioReagents™
BP2558-10 |
PhiX-174 RF DNAHae III Digest | Liquid | 10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Compatible with both conventional and pulsed field gel electrophoresis
- DNase-, RNase-, and protease-free
Specifications
Colorless | |
DNA Molecular Weight Marker/Ladder | |
Determining the size of DNA fragment |