missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Microbiology Media: LB Broth, Miller

Used for growing molecular biology strains of E. coli-containing recombinant plasmids

Brand:  Fisher BioReagents™ BP1426-2

Code : 32

Additional Details : Weight : 2.15448kg

 View more versions of this product

Product Code. 10638013

  • £176.13 / 2kg


Estimated Shipment
07-06-2018
Add to basket

Description

Description

Dehydrated.

Specifications

Specifications

Bacterial Culture
Powder
LB Broth
2000g
LB broth, Miller
Molecular genetics
Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L
Poly Pail
4°C
For preparing bacterial nutrient media
Videos
SDS Documentation